missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mometasone furoate

Catalog No. ac462130010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462130010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC462130010 Supplier Thermo Scientific Chemicals Supplier No. 462130010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83919-23-7
Molecular Formula C27H30Cl2O6
Molecular Weight (g/mol) 521.43
InChI Key WOFMFGQZHJDGCX-ZULDAHANSA-N
PubChem CID 126963492

Specifications

Melting Point 218°C to 220°C
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,methanol
Formula Weight 521.43
Chemical Name or Material Mometasone furoate
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only