missing translation for 'onlineSavingsMsg'
Learn More

Solstice Molybdic Acid, Solstice

Catalog No. 6002165
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002165 500 g
6002164 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002165 Supplier Solstice Supplier No. 232084500G

missing translation for 'validMainframeMsg'

ACS Reagent,≥85.0% MoO3 basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7782-91-4
Molecular Formula H2MoO4
Molecular Weight (g/mol) 161.96
MDL Number MFCD00036287
InChI Key VLAPMBHFAWRUQP-UHFFFAOYSA-L
Synonym molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2
PubChem CID 82208
ChEBI CHEBI:25371
IUPAC Name dihydroxy(dioxo)molybdenum
SMILES O[Mo](O)(=O)=O

Specifications

Physical Form Solid
Quantity 500 g
Assay Percent Range 85.0% min
Density 3.1g/mL at 25°C; Vapor Density: 6.2 (vs air)
Linear Formula H2MoO4
UN Number NONH for all modes of transport
Formula Weight 161.96 g/mol
Percent Purity ≥85% (MoO3 basis)
Grade ACS Reagent
Assay MoO3 basis
Chemical Name or Material Molybdic Acid, ACS Reagent, ≥85% (MoO3 basis)
Show More Show Less
Hazard Statement P261-P305 + P351 + P338
Precautionary Statement H319-H335-H373
EINECSNumber 231-970-5