missing translation for 'onlineSavingsMsg'
Learn More

Molybdic acid, for analysis

CAS: 7782-91-4 | H2MoO4 | 161.96 g/mol

$148.45 - $148.45

Chemical Identifiers

CAS 7782-91-4
Molecular Formula H2MoO4
Molecular Weight (g/mol) 161.96
MDL Number MFCD00036287
InChI Key VLAPMBHFAWRUQP-UHFFFAOYSA-L
Synonym molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2
PubChem CID 82208
ChEBI CHEBI:25371
IUPAC Name dihydroxy(dioxo)molybdenum
SMILES O[Mo](O)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC213621000
SDS View Documents
Thermo Scientific Chemicals
213621000
100 g Plastic bottle
Each for $148.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

7782-91-4
161.96
VLAPMBHFAWRUQP-UHFFFAOYSA-L
82208
dihydroxy(dioxo)molybdenum
H2MoO4
MFCD00036287
molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2
CHEBI:25371
O[Mo](O)(=O)=O

Specifications

7782-91-4
Powder
H2MoO4
15, 6326
Solubility in water: insoluble. Other solubilities: soluble in nh4oh,h2so4 and fixed alkalies
O[Mo](O)(=O)=O
161.96
CHEBI:25371
≥85% (MoO3)
Plastic bottle
3.1000g/mL
White through Yellow
100 g
MFCD00036287
molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2
VLAPMBHFAWRUQP-UHFFFAOYSA-L
dihydroxy(dioxo)molybdenum
82208
161.96
Analytical
3.1
Molybdic acid, p.a.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.

GHS P Statement
Wear eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/

GHS Signal Word: Warning

EINECSNumber : 231-970-5

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only