missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdic acid, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline Powder and/or Crystals |
| Quantity | 100 g |
| Packaging | Plastic bottle |
| Specific Gravity | 3.1 |
| Density | 3.1000g/mL |
| Merck Index | 15,6326 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in nh4oh, h2so4 and fixed alkalies |
| Formula Weight | 161.96 |
| Percent Purity | ≥85.0% (MoO3) |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. May cause damage to organs through prolonged or repeated exposure. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear eye protection/face protection. If eye irritation persists: Get medical advice/attention. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/ |
| EINECSNumber | 231-970-5 |
RUO – Research Use Only