missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Molybdenum(VI) sulfide, 97%
CAS: 12033-29-3 | MoS{3}
$72.90 - $180.05
Chemical Identifiers
| CAS | 12033-29-3 |
|---|---|
| Molecular Formula | MoS{3} |
| MDL Number | MFCD00014226 |
Chemical Identifiers
| 12033-29-3 | |
| MoS{3} | |
| MFCD00014226 |
Specifications
| 12033-29-3 | |
| 25 g | |
| MFCD00014226 | |
| 97% |
| Solid | |
| MoS{3} | |
| 192.13 |
Safety and Handling
ShelfLife : 5 Years