missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(V) isopropoxide, 99+% (metals basis)

Catalog No. p-7051540
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4148402 0.5 g
AA4148404 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4148402 Supplier Thermo Scientific Chemicals Supplier No. 04148402
Only null left

Precursor used in sol-gel process | CAS: 209733-38-0 | C15H35MoO5 | 391.39 g/mol

Molybdenum(V) isopropoxide, it is used as a Precursor used in sol-gel process. It is used in nanomaterials and thin films. It is used as a solution based product for printed electronics. It acts a catalyst for selective isobutane oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 209733-38-0
Molecular Formula C15H35MoO5
Molecular Weight (g/mol) 391.39
MDL Number MFCD00210636
InChI Key HLFWWCCBFJUYJL-UHFFFAOYSA-N
Synonym molybdenum v isopropoxide, acmc-20akdz, pentaisopropoxymolybdenum v, molybdenum 5+ pentakis propan-2-olate
PubChem CID 18475294
IUPAC Name molybdenum;propan-2-olate
SMILES CC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C

Specifications

Color Black to Yellow
Linear Formula Mo[OCH(CH3)2]5
Quantity 0.5 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information It is soluble in hydrocarbons,reacts with alcohols,ketones and esters. It reacts exothermally with water.
Formula Weight 391.04
Percent Purity ≥99%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Molybdenum(V) isopropoxide
Show More Show Less
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only