missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum(V) isopropoxide, 99.6% (m.b.), 5% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles
Description
Used in the preparations of nanomaterials and thin films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Specifications
| Color | Yellow to Brown |
| Flash Point | 12°C (54°F) |
| Odor | Alcohol-like |
| Packaging | ChemSeal™ bottles |
| Linear Formula | Mo[OCH(CH3)2]5 |
| Quantity | 25 mL |
| UN Number | UN1219 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in hydrocarbons,reacts with alcohols,ketones and esters |
| Formula Weight | 391.04 |
| Show More |
Safety and Handling
| Hazard Category | H225-H319-H335-H336 |
| Hazard Statement | GHS H Statement H225-H315-H319-H336 Highly flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL |
| TSCA | No |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only