missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(V) isopropoxide, 99.6% (m.b.), 5% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AA41836AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41836AC 25 mL
AA41836AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41836AC Supplier Thermo Scientific Chemicals Supplier No. 041836AC

CAS: 209733-38-0 | C15H35MoO5 | 391.39 g/mol

Used in the preparations of nanomaterials and thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon

Chemical Identifiers

CAS 209733-38-0
Molecular Formula C15H35MoO5
Molecular Weight (g/mol) 391.39
MDL Number MFCD00210636
InChI Key HLFWWCCBFJUYJL-UHFFFAOYSA-N
Synonym molybdenum v isopropoxide, acmc-20akdz, pentaisopropoxymolybdenum v, molybdenum 5+ pentakis propan-2-olate
PubChem CID 18475294
IUPAC Name molybdenum;propan-2-olate
SMILES CC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C

Specifications

Color Yellow to Brown
Flash Point 12°C (54°F)
Odor Alcohol-like
Packaging ChemSeal™ bottles
Linear Formula Mo[OCH(CH3)2]5
Quantity 25 mL
UN Number UN1219
Sensitivity Moisture sensitive
Solubility Information Soluble in hydrocarbons,reacts with alcohols,ketones and esters
Formula Weight 391.04
Percent Purity 99.6%
Concentration or Composition (by Analyte or Components) 5% w/v in isopropanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Molybdenum(V) isopropoxide
Show More Show Less
Hazard Category H225-H319-H335-H336
Hazard Statement GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
TSCA No
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only