missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum rod, 6.4mm (0.25in) dia, 99.95% (metals basis)

Catalog No. p-4824821
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1003318 50 g
AA1003330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1003318 Supplier Thermo Scientific Chemicals Supplier No. 01003318
Only null left

Approx. 15.2cm/50g | CAS: 7439-98-7 | Mo | 95.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Color Gray
Appearance Dark gray rod
Quantity 50 g
Odor Odorless
Form Rod
Assay Percent Range 99.95% (Metals basis)
Chemical Name or Material Molybdenum
EINECSNumber 231-107-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only