missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum rod, 3.17mm (0.125in) dia, annealed, 99.95% (metals basis)

Catalog No. p-7048282
Change view
Click to view available options
Quantity:
50 cm
5 x 50 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10032BW 50 cm
AA10032VP 5 x 50 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA10032BW Supplier Thermo Scientific Chemicals Supplier No. 010032BW
Only null left

CAS: 7439-98-7 | Mo | 95.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Quantity 50 cm
Weight ≈0.72g/cm
Form Rod
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Molybdenum rod, 3.17mm (0.125 in.) dia., Annealed
EINECSNumber 231-107-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only