missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum powder, spherical, -170 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7051760
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4191918 50 g
AA4191930 250 g
AA41919A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4191918 Supplier Thermo Scientific Chemicals Supplier No. 04191918
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Color Gray
Quantity 50 g
Mesh Size −170 Mesh
Odor Odorless
Form Spherical Powder
UN Number UN3089
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Molybdenum powder
Hazard Category H228-H360-H371
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P201-P202-P210-P240-P241-P260-P264b-P270-P281-P308+P313-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 231-107-2
RTECSNumber QA4680000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only