missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum powder, APS typically 2-4 micron, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog No. p-4824819
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3968714 25 g
AA3968722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3968714 Supplier Thermo Scientific Chemicals Supplier No. 03968714
Only null left

From Hydrogen reduced oxide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Color Gray
Appearance gray powder
Quantity 25 g
Odor Odorless
Form Powder
Assay Percent Range 99.9% (Metals basis)
Chemical Name or Material Molybdenum
Hazard Category H228-H360-H371
Precautionary Statement P201-P202-P210-P240-P241-P260-P264b-P270-P281-P308+P313-P370+P378q-P501c
DOTInformation DOT Class: 4.1; Packaging Group: II
EINECSNumber 231-107-2
RTECSNumber QA4680000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only