missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum nanopowder, APS 60-80nm, 99% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7059952
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4579406 5 g
AA4579414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4579406 Supplier Thermo Scientific Chemicals Supplier No. 04579406
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Quantity 5 g
Odor Odorless
Particle Size APS 60 to 80nm
Form Nanopowder
UN Number UN3089
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Molybdenum nanopowder
Hazard Category H228-H360-H371
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P201-P202-P210-P240-P241-P260-P264b-P270-P281-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
EINECSNumber 231-107-2
RTECSNumber QA4680000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only