Learn More
Molybdenum(IV) sulfide, 98%
CAS: 1317-33-5 | MoS2 | 160.07 g/mol
$51.90 - $427.21
Chemical Identifiers
| CAS | 1317-33-5 |
|---|---|
| Molecular Formula | MoS2 |
| Molecular Weight (g/mol) | 160.07 |
| MDL Number | MFCD00003470 |
| InChI Key | CWQXQMHSOZUFJS-UHFFFAOYSA-N |
| Synonym | molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z |
| PubChem CID | 14823 |
| ChEBI | CHEBI:30704 |
| SMILES | S=[Mo]=S |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4182714
|
Thermo Scientific Chemicals
04182714 |
25 g |
Each for $51.90
|
|
|||||
|
AA4182722
|
Thermo Scientific Chemicals
04182722 |
100 g |
Each for $134.33
|
|
|||||
|
AA4182736
|
Thermo Scientific Chemicals
04182736 |
500 g |
Each for $427.21
|
|
|||||
Description
Molybdenum(IV) sulfide is used as a catalyst in the electrolysis of water. It is employed as a cocatalyst for desulfurization in petrochemistry especially hydrodesulfurization. It is also used as a component of photoelectrochemical and microelectronics applications. Further, it is useful as a filament material in many electrical applications. It finds application as a lubricant in aircraft engines. Polymers consisting of molybdenum(IV) sulfide such as nylon, teflon and vespel posses improved strength and reduced friction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1317-33-5 | |
| 160.07 | |
| CWQXQMHSOZUFJS-UHFFFAOYSA-N | |
| 14823 | |
| S=[Mo]=S |
| MoS2 | |
| MFCD00003470 | |
| molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z | |
| CHEBI:30704 |
Specifications
| 1185°C | |
| Black | |
| 25 g | |
| MoS2 | |
| molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z | |
| CWQXQMHSOZUFJS-UHFFFAOYSA-N | |
| disulfanylidenemolybdenum | |
| 14823 | |
| 160.08 | |
| Black powder |
| 1317-33-5 | |
| Powder | |
| 99% (metals basis) | |
| MFCD00003470 | |
| Soluble in hot sulfuric acid,and aquaregia. Insoluble in water,concentrated sulfuric acid and dilute acid. | |
| S=[Mo]=S | |
| 160.07 | |
| CHEBI:30704 | |
| Odorless | |
| Molybdenum(IV) sulfide |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
P261-P271-P304+P340-P312
H332
EINECSNumber : 215-263-9
RTECSNumber : QA4697000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only