missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(IV) sulfide, 98%

Catalog No. p-4782005
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1221330 250 g
AA12213A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1221330 Supplier Thermo Scientific Chemicals Supplier No. 01221330

Hydrogenation and Isomerization Catalyst, Dry Lubricant | CAS: 1317-33-5 | MoS2 | 160.07 g/mol

It is widely used as a solid lubricant .It finds applications mainly as Intermediates, Process regulators, Processing aids. Employed as a co catalyst for desulfurization in petrochemistry. As a catalyst in the electrolysis of water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1317-33-5
Molecular Formula MoS2
Molecular Weight (g/mol) 160.07
MDL Number MFCD00003470
InChI Key CWQXQMHSOZUFJS-UHFFFAOYSA-N
Synonym molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z
PubChem CID 14823
ChEBI CHEBI:30704
SMILES S=[Mo]=S

Specifications

Melting Point 1185°C
Color Black
Physical Form Powder
Quantity 250 g
Assay Percent Range 98%
Solubility Information Insoluble in water and dilute acids
IUPAC Name disulfanylidenemolybdenum
Formula Weight 160.08
Odor Odorless
Appearance Black powder
Chemical Name or Material Molybdenum(IV) sulfide
Show More Show Less
Hazard Category H332
Precautionary Statement P261-P271-P304+P340-P312
EINECSNumber 215-263-9
RTECSNumber QA4697000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only