missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum(IV) sulfide, 98.5%
CAS: 1317-33-5 | MoS2 | 160.07 g/mol
$128.17 - $431.40
Chemical Identifiers
| CAS | 1317-33-5 |
|---|---|
| Molecular Formula | MoS2 |
| Molecular Weight (g/mol) | 160.07 |
| InChI Key | CWQXQMHSOZUFJS-UHFFFAOYSA-N |
| Synonym | molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z |
| PubChem CID | 14823 |
| ChEBI | CHEBI:30704 |
| IUPAC Name | bis(sulfanylidene)molybdenum |
| SMILES | S=[Mo]=S |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC215781000
|
Thermo Scientific Chemicals
215781000 |
100 g | Glass bottle |
Each for $128.17
|
|
||||
|
AC215785000
|
Thermo Scientific Chemicals
215785000 |
500 g | Glass bottle |
Each for $431.40
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1317-33-5 | |
| 160.07 | |
| molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z | |
| CHEBI:30704 | |
| S=[Mo]=S |
| MoS2 | |
| CWQXQMHSOZUFJS-UHFFFAOYSA-N | |
| 14823 | |
| bis(sulfanylidene)molybdenum |
Specifications
| 2375.0°C | |
| 1317-33-5 | |
| 100 g | |
| MoS2 | |
| 15,6322 | |
| Solubility in water: insoluble. Other solubilities: soluble in aqua regia and sulfuric acid | |
| S=[Mo]=S | |
| 160.07 | |
| CHEBI:30704 | |
| 98.5% | |
| Molybdenum(IV) sulfide |
| Black or Gray | |
| Powder | |
| 98% min. (MoS2) | |
| MoS2 | |
| molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z | |
| CWQXQMHSOZUFJS-UHFFFAOYSA-N | |
| bis(sulfanylidene)molybdenum | |
| 14823 | |
| 160.07 | |
| Glass bottle |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 215-263-9
RUO – Research Use Only