missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum gauze, 100 mesh woven from 0.0636mm (0.0025in) dia wire
CAS: 7439-98-7 | Mo | 95.95 g/mol
Supplier: Thermo Scientific Chemicals 040912GT
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Molybdenum gauze | |
| 7439-98-7 | |
| 100 Mesh | |
| Mo | |
| molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 | |
| [Mo] | |
| 95.95 | |
| CHEBI:28685 |
| Woven from 0.0636mm (0.0025 in.) dia. wire | |
| 150 x 150 mm | |
| Wire Cloth | |
| MFCD00003465 | |
| ZOKXTWBITQBERF-UHFFFAOYSA-N | |
| molybdenum | |
| 23932 |
Chemical Identifiers
| 7439-98-7 | |
| 95.95 | |
| ZOKXTWBITQBERF-UHFFFAOYSA-N | |
| 23932 | |
| molybdenum |
| Mo | |
| MFCD00003465 | |
| molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 | |
| CHEBI:28685 | |
| [Mo] |
Safety and Handling
EINECSNumber : 231-107-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only