missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum gauze, 100 mesh woven from 0.0636mm (0.0025in) dia wire

Catalog No. AA40912FL
Change view
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40912FL 75 x 75 mm
AA40912GT 150 x 150 mm
AA40912HB 300 x 300 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40912FL Supplier Thermo Scientific Chemicals Supplier No. 040912FL

CAS: 7439-98-7 | Mo | 95.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Quantity 75 x 75 mm
Mesh Size 100 Mesh
Form Wire Cloth
Chemical Name or Material Molybdenum gauze, Woven from 0.0636mm (0.0025 in.) dia. wire
EINECSNumber 231-107-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only