missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum foil, 1.0mm (0.04in) thick, 99.95% (metals basis)

Catalog No. p-7048287
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 x 100 mm
200 x 200 mm
50 x 50 mm
Catalog No. Quantity
AA10046FI 50 x 50 mm
AA10046GH 100 x 100 mm
AA10046GW 200 x 200 mm
3 options

Catalog No. AA10046FI

Supplier: Thermo Scientific Chemicals 010046FI

Only null left

CAS: 7439-98-7 | Mo | 95.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Quantity 50 x 50 mm
Weight ≈25.6g/50x50mm
Form Foil
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Molybdenum foil, 1.0mm (0.04 in.) thick
EINECSNumber 231-107-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only