missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum foil, 0.127mm (0.005in) thick, 99.95% (metals basis)

Catalog No. p-7051684
Change view
Click to view available options
Quantity:
100 x 150 mm
150 x 300 mm
300 x 600 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41764GJ 100 x 150 mm
AA41764HW 150 x 300 mm
AA41764HX 300 x 600 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41764GJ Supplier Thermo Scientific Chemicals Supplier No. 041764GJ
Only null left

CAS: 7439-98-7 | Mo | 95.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Quantity 100 x 150 mm
Weight ≈19.5g/100x150mm
Form Foil
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Molybdenum foil, 0.127mm (0.005 in.) thick, Hard
EINECSNumber 231-107-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only