missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum foil, 0.025mm (0.001in) thick, 99.95% (metals basis)

Catalog No. p-4782000
Change view
Click to view available options
Quantity:
150 x 150 mm
300 x 300 mm
300 x 1200 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10042GT 150 x 150 mm
AA10042HB 300 x 300 mm
AA10042HZ 300 x 1200 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA10042GT Supplier Thermo Scientific Chemicals Supplier No. 010042GT
Only null left

Approx. 5.8g/150 x 150mm | CAS: 7439-98-7 | Mo | 95.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Color Gray
Appearance Dark gray foil
Density 10.22g/cm3 at 20°C
Quantity 150 x 150 mm
Odor Odorless
Form Foil
Assay Percent Range 99.95% (Metals basis)
Chemical Name or Material Molybdenum
EINECSNumber 231-107-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only