missing translation for 'onlineSavingsMsg'
Learn More

Molsidomine 98.0+%, TCI America™

Catalog No. M25241G
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
M25241G 1 g
M25245G 5 g
2 options

Catalog No. M25241G

Supplier: TCI America M25241G

Only null left

Chemical Identifiers

CAS 25717-80-0
Molecular Formula C9H14N4O4
Molecular Weight (g/mol) 242.235
MDL Number MFCD00869301
InChI Key XLFWDASMENKTKL-UHFFFAOYSA-N
PubChem CID 5353788
IUPAC Name (1E)-1-ethoxy-N-(3-morpholin-4-yloxadiazol-3-ium-5-yl)methanimidate
SMILES CCOC(=NC1=C[N+](=NO1)N2CCOCC2)[O-]

Specifications

Melting Point 141°C
Color White
Quantity 1 g
Formula Weight 242.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Molsidomine
EINECSNumber (9)-2224
RTECSNumber WU7677400
TSCA No
Recommended Storage Refrigerator