missing translation for 'onlineSavingsMsg'
Learn More

Molindone Hydrochloride 98.0+%, TCI America™

Catalog No. M22521G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22521G 1 g
M2252100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22521G Supplier TCI America Supplier No. M22521G

Chemical Identifiers

CAS 15622-65-8
Molecular Formula C16H25ClN2O2
Molecular Weight (g/mol) 312.84
MDL Number MFCD01718304
InChI Key GQWNECFJGBQMBO-UHFFFAOYNA-N
Synonym 3-Ethyl-6,7-dihydro-2-methyl-5-(morpholinomethyl)-1H-indol-4(5H)-one Hydrochloride, 3-Ethyl-1,5,6,7-tetrahydro-2-methyl-5-(4-morpholinylmethyl)-4H-indol-4-one Hydrochloride
PubChem CID 9883259
IUPAC Name 4-[(3-ethyl-2-methyl-4-oxo-4,5,6,7-tetrahydro-1H-indol-5-yl)methyl]morpholin-4-ium chloride
SMILES [Cl-].CCC1=C(C)NC2=C1C(=O)C(C[NH+]1CCOCC1)CC2

Specifications

Quantity 1 g
UN Number 2811
Formula Weight 312.84
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Molindone Hydrochloride
RTECSNumber NM3325000
TSCA No