missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNP-ATP (2'-(or-3')-O-(Trinitrophenyl) Adenosine 5'-Triphosphate, Trisodium Salt)

Catalog No. t7602
Encompass
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T7602 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T7602 Supplier Invitrogen™ Supplier No. T7602

Structural probe

The nucleotide analog TNP-ATP is modified on the ribose moiety and is essentially non-fluorescent in water. The TNP nucleotides undergo an equilibrium transition to a semiquinoid structure that has relatively long-wavelength spectral properties; this form is only fluorescent when bound to the nucleotide-binding site of some proteins.

Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation

Order Info

Shipping Condition: Wet ice

Specifications

Product Type TNP-ATP
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Shipping Condition Wet Ice
Label or Dye TNP
Quantity 2 mL
What is the concentration of TNP-ATP (2'-(or-3')-O-(Trinitrophenyl) Adenosine 5'-Triphosphate, Trisodium Salt) (Cat. No. T7602)?

The concentration is 5 mg/mL. This information can also be found on the product COA.

For Research Use Only. Not for use in diagnostic procedures.