missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TC-ReAsH™ II In-Cell Tetracysteine Tag Detection Kit (Red Fluorescence), for live-cell imaging

Catalog No. T34562
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34562 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T34562 Supplier Invitrogen™ Supplier No. T34562
Only null left

Includes

Kit contains ReAsH-EDT2 labeling reagent and BAL wash buffer.

Contains an expression tag-based fluorescence labeling reagent for live-cell labeling

Mammalian cell lines expressing a protein fused to a tetracysteine tag (CCPGCC) can be labeled with the red-fluorescent ReAsH-EDT2 reagent contained in the kit. The tagged protein is fluorescent only when the labeling reagent is added. BAL wash buffer replaces the previously supplied Disperse Blue 3 and EDT-based wash buffer as an olfactorily more agreeable reagent that yields superior signal to noise.

Cell Analysis, Cell-Based Reporter Assays, Cellular Imaging, Enzyme and Protein Activity Assays, Labeling Chemistry, Lumio Detection, Proteins, Expression, Isolation and Analysis, Reporter Assays, Tetracysteine (Lumio/FlAsH/ReAsH) Assays

Order Info

Shipping Condition: Wet Ice

Specifications

Product Type Tetracysteine Tag Detection Kit
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Content And Storage Store in freezer (-5 to -30°C).
Color Red
Label or Dye ReAsH
Shipping Condition Wet Ice
Format Kit
Product Line TC-ReAsH II
Quantity 1 Kit

For Research Use Only. Not for use in diagnostic procedures.