missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tali™ Cell Cycle Kit

Catalog No. a10798
Encompass
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
50 Assays
Catalog No. Quantity
A10798 50 Assays
1 options

Catalog No. A10798

Supplier: Invitrogen™ A10798

Only null left

Includes

1 bottle

Convenient and accurate determination of the percentage of cells in each phase of the cell cycle

The TaliCell Cycle Kit includes a ready-to-use reagent that can be used with the TaliImage-Based Cytometer. This single-addition reagent is composed of propidium iodide (PI), RNase A, and Triton X-100.

The TaliCell Cycle Kit offers a simple workflow:

  • Pre-mixed reagent can be stored at room temperature and used when your cells are ready
  • Simply fix your cells, add the reagent, incubate, and visualize
  • Cell cycle data can be analyzed on the Taliinstrument or exported as a .csv or .fcs file for analysis with various modeling software
Fluorescent detection of cellular DNA as a cell-cycle phase indicator

Cell Analysis, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Cellular Imaging, DNA Replication and Repair, General Cell Tracing, Imaging Cytometry

Specifications

Product Type Cell Cycle Kit
Cell Type Mammalian Cell
Detection Method Fluorescence
For Use With (Equipment) Tali™ Image-Based Cytometer, Tali™ Image-Based Cytomoter
Content And Storage 1 bottle, store at room temperature.
Color Red
Compatible Cells Mammalian Cells
Label or Dye Propidium Iodide
Format Bottle
Product Line Tali
Quantity 50 Assays
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.