missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYPRO™ Protein Gel Stains

Catalog No. p-4918382
Encompass
Change view
Click to view available options
Color:
Orange
Red
Ruby
Tangerine
Quantity:
500 μL
10 x 50 μL
200 mL
1 L
5 L
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
S6650 Orange 500 μL
S6651 Orange 10 x 50 μL
S12010 Tangerine 500 μL
S6653 Red 500 μL
S6654 Red 10 x 50 μL
S12001 Ruby 200 mL
S12000 Ruby 1 L
S21900 Ruby 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. S6650 Supplier Invitrogen™ Supplier No. S6650
Only null left

SYPRO protein gel stains are sensitive, easy-to-use fluorescent stains for total proteins separated by polyacrylamide gel electrophoresis (PAGE).

SYPRO protein gel stains are sensitive, easy-to-use fluorescent stains for total proteins separated by polyacrylamide gel electrophoresis (PAGE). Stained proteins can be viewed with a standard UV or blue-light transilluminator, or imaging equipment containing the appropriate filters or laser. SYPRO protein gel stains offer many advantages over Coomassie Blue and silver staining, including a fast, one-step staining protocol requiring no destaining, a linear dynamic range over three orders of magnitude, and very little protein-to-protein variation in staining.

SYPRO Ruby Protein Gel Stain features include:

  • Convenient—ready-to-use formulation
  • High sensitivity—detection to 0.25 to 1 ng
  • Effective staining for 1D and 2D gels
  • Can be multiplexed with other gel stains such as Pro-Q Diamond phosphoprotein gel stain or Pro-Q Emerald 300 glycoprotein gel stain
  • Compatible with subsequent analysis of proteins by Edman-based sequencing or mass spectrometry

SYPRO Orange and SYPRO Red protein gel stain features include:

  • Sensitive—detection to 4 to 8 ng
  • Fast—rapid staining time of 10–60 min
  • Best for 1D gels
  • SYPRO Orange: Ex: 300, 470 nm, Em: 570 nm
  • SYPRO Red: Ex: 300, 550 nm, Em: 630 nm

SYPRO Tangerine Protein Gel Stain features include:

  • Sensitive—detection to 4 to 8 ng
  • Fast—rapid staining time of 10 to 60 minutes
  • Best for 1D gels
  • Does not require fixation with acetic acid
  • Compatible with subsequent gel staining, western blotting, zymography, electroelution or mass spectrometry

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Supplied as a 5000X concentrate in DMSO. Store at room temperature and protect from light.
Detection Location In-Gel Detection
Color Orange
Concentration 5000X in DMSO
Detection Method Fluorescence
Label or Dye SYPRO Orange
Quantity 500 μL
Shipping Condition Room Temperature
Target Molecule Protein
Excitation/Emission 300, 470/570 nm
Product Line SYPRO
Product Type Protein Gel Stain
Show More Show Less