missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYPRO™ Protein Gel Stains

Catalog No. S6654
Encompass
Change view
Click to view available options
Color:
Orange
Red
Ruby
Tangerine
Quantity:
500 μL
10 x 50 μL
200 mL
1 L
5 L
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
S6654 Red 10 x 50 μL
S6650 Orange 500 μL
S6651 Orange 10 x 50 μL
S12010 Tangerine 500 μL
S6653 Red 500 μL
S12001 Ruby 200 mL
S12000 Ruby 1 L
S21900 Ruby 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. S6654 Supplier Invitrogen™ Supplier No. S6654
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SYPRO protein gel stains are sensitive, easy-to-use fluorescent stains for total proteins separated by polyacrylamide gel electrophoresis (PAGE).

SYPRO protein gel stains are sensitive, easy-to-use fluorescent stains for total proteins separated by polyacrylamide gel electrophoresis (PAGE). Stained proteins can be viewed with a standard UV or blue-light transilluminator, or imaging equipment containing the appropriate filters or laser. SYPRO protein gel stains offer many advantages over Coomassie Blue and silver staining, including a fast, one-step staining protocol requiring no destaining, a linear dynamic range over three orders of magnitude, and very little protein-to-protein variation in staining.

SYPRO Ruby Protein Gel Stain features include:

  • Convenient—ready-to-use formulation
  • High sensitivity—detection to 0.25 to 1 ng
  • Effective staining for 1D and 2D gels
  • Can be multiplexed with other gel stains such as Pro-Q Diamond phosphoprotein gel stain or Pro-Q Emerald 300 glycoprotein gel stain
  • Compatible with subsequent analysis of proteins by Edman-based sequencing or mass spectrometry

SYPRO Orange and SYPRO Red protein gel stain features include:

  • Sensitive—detection to 4 to 8 ng
  • Fast—rapid staining time of 10–60 min
  • Best for 1D gels
  • SYPRO Orange: Ex: 300, 470 nm, Em: 570 nm
  • SYPRO Red: Ex: 300, 550 nm, Em: 630 nm

SYPRO Tangerine Protein Gel Stain features include:

  • Sensitive—detection to 4 to 8 ng
  • Fast—rapid staining time of 10 to 60 minutes
  • Best for 1D gels
  • Does not require fixation with acetic acid
  • Compatible with subsequent gel staining, western blotting, zymography, electroelution or mass spectrometry

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store at room temperature and protect from light.
Detection Location In-Gel Detection
Color Red
Concentration 5000X in DMSO
Detection Method Fluorescence
Label or Dye SYPRO Red
Quantity 10 x 50 μL
Shipping Condition Room Temperature
Target Molecule Protein
Excitation/Emission 300, 550/630 nm
Product Line SYPRO
Product Type Protein Gel Stain
Show More Show Less
If I increase the final concentration of my SYPRO Orange or SYPRO Red staining solution above 1X, can I increase the signal of my stained proteins?

No. Dye concentrations higher than 1X do not give better detection. Instead, background fluorescence increases and, as the dye concentration increases, the dye becomes self-quenching and the signal actually decreases.

Can I dry my SYPRO Ruby, SYPRO Orange, or SYPRO Red stained gels?

Gels stained with SYPRO dyes can be dried between sheets of cellophane, although there is sometimes a slight decrease in sensitivity. If the gels are dried onto paper, the light will scatter and the sensitivity will decrease. Other plastics are not recommended, as the plastic typically used is not transparent to UV light.

Can I pre-stain proteins with SYPRO Ruby, SYPRO Orange or SYPRO Red Protein Gel Stains and then run them through a gel?

No. Loading solutions contain so much SDS that SYPRO Ruby, SYPRO Orange and SYPRO Red dyes simply localize in the free SDS and bind very little of the proteins. Proteins can be covalently pre-labeled with ATTO-TAG CBQCA (Cat. No. A2333), DDAO succinimidyl ester (Cat. No. C34553) or TAMRA-succinimidyl ester (Cat. No. C2211) dyes, or the TC-FLAsH Expression Analysis Detection Kits (Cat. No. A10067 for orange fluorescence, Cat. No. A10068 for red fluorescence) prior to electrophoresis without affecting protein migration through the gel.

SYPRO Orange or SYPRO Red Protein Gel Stain can be diluted 5000-fold into the cathode (upper) buffer tank to stain proteins during electrophoresis without affecting migration. The problem with doing this is that there is considerable background fluorescence in the gels from the dye interacting with SDS. This can be reduced after electrophoresis by destaining in 7.5% acetic acid for 15-60 minutes. This method also results in poorer protein sensitivity than the standard post-staining method, requires the same amount of time before the gel can be imaged, and contaminates the electrophoresis apparatus.