missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Electrophoretic Mobility-Shift Assay (EMSA) Kit, with SYBR™ Green & SYPRO™ Ruby EMSA stains

Catalog No. e33075
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E33075 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E33075 Supplier Invitrogen™ Supplier No. E33075
Only null left

Detects both nucleic acid and protein in the same gel

This fluorescence-based Electrophoretic Mobility Shift Assay (EMSA) Kit provides a fast, easy and quantitative method. This kit uses two fluorescent dyes for detection - SYBR™ Green EMSA nucleic acid gel stain for RNA or DNA detection and SYPRO™ Ruby EMSA protein gel stain for protein detection. The nucleic acids and proteins are stained sequentially in the gel after electrophoresis, so there is no possibility that labeling will interfere with the protein binding being assayed.

Enzyme and Protein Activity Assays, Proteins, Expression, Isolation and Analysis, RNAi, Epigenetics and Non-Coding RNA Research, Transcription Factor Activity and Detection

Order Info

Shipping Condition: Room Temperature

Specifications

Assay EMSA (Electrophoretic Mobility Shift Assay)
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Quantity 1 Kit
Shipping Condition Room Temperature
Product Line SYBR, SYPRO
Product Type Electrophoretic Mobility-Shift Assay (EMSA) Kit

For Research Use Only. Not for use in diagnostic procedures.