missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Secure-Seal™ Spacer, one well, 13 mm diameter, 0.12 mm deep

Catalog No. S24735
Encompass
Change view
Click to view available options
Quantity:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S24735 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24735 Supplier Invitrogen™ Supplier No. S24735
  • ONLINE EXCLUSIVE PRICE
    $340.00
    $278.80 / Each

    Save $61.20

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Removable hydrophobic barrier that provides flexibility for isolating specimens

  • Barriers may be used to isolate cells grown in culture dishes or to separate specimens on microscopy slides during staining procedures
  • Silicone material is autoclavable and adheres to any smooth surface

Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunohistochemistry (IHC)

Order Info

Shipping Condition: Room temperature

Specifications

Type Spacer
Product Line Secure-Seal
Depth (Metric) 0.12 mm
Diameter (Metric) 13 mm
Content And Storage Store at room temperature.
Quantity 1 set
Shipping Condition Room Temperature
I would like to know what quantity corresponds to 1 set of Secure-Seal Spacer, one well, 13 mm diameter, 0.12 mm deep (Cat. No. S24735)?

It contains 100 spacers.

For Research Use Only. Not for use in diagnostic procedures.