missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RH 237 (N-(4-Sulfobutyl)-4-(6-(4-(Dibutylamino)phenyl)hexatrienyl)Pyridinium, Inner Salt)

Catalog No. S1109
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S1109 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S1109 Supplier Invitrogen™ Supplier No. S1109
  • ONLINE EXCLUSIVE PRICE
    $708.00
    $580.56 / Each

    Save $127.44

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fast-response potential-sensitive probe

RH 237 can be used for imaging functional neurons.

Cell Analysis, Cell Metabolism, Cell Structure, Cell Viability, Proliferation & Function, Membranes (General) & Lipids

Order Info

Shipping Condition: Room Temperature

Specifications

Color Infrared
Detection Method Fluorescence
Product Type Sensitive Probe
Quantity 5 mg
Content And Storage Store at room temperature and protect from light.
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope
I am seeing high background outside of my neuronal cells when using membrane potential indicators. What can I do to reduce background?

If you use our FluoVolt Membrane Potential Kit (Cat. No. F10488), the kit provides a background suppressor to reduce this problem. For other indicators, consider the use of BackDrop Background Suppressor (Cat no. R37603, B10511, and B10512).

What is the difference between fast and slow-response membrane potential probes?

Molecules that change their structure in response to the surrounding electric field can function as fast-response probes for the detection of transient (millisecond) potential changes. Slow-response dyes function by entering depolarized cells and binding to proteins or membranes. Increased depolarization results in additional dye influx and an increase in fluorescence, while hyperpolarization is indicated by a decrease in fluorescence. Fast-response probes are commonly used to image electrical activity from intact heart tissues or measure membrane potential changes in response to pharmacological stimuli. Slow-responding probes are often used to explore mitochondrial function and cell viability.

What type of membrane potential indicators do you offer and how should I choose one for my experiment?

A membrane potential indicator selection guide can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-viability-and-regulation/ion-indicators/membrane-potential-indicators.html).

For Research Use Only. Not for use in diagnostic procedures.