missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PS-Speck™ Microscope Point Source Kit (blue, green, orange & deep-red fluorescent beads)

Catalog No. P7220
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P7220 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P7220 Supplier Invitrogen™ Supplier No. P7220

Includes

Mounting protocol and mounting medium to prepare 100 slides

Contains four different fluorescence microsphere colors

Each microsphere has a diameter of 0.175 ±0.005μm. This exceptionally small diameter coefficient of variation makes the PS-Speck microspheres ideal as uniform, subresolution fluorescent point sources for calibrating instrument optics, especially in three-dimensional imaging applications.

  • Four ready-to-use 1mL suspensions contain bright, monodisperse microspheres with excitation/emission wavelengths of 360/440nm (blue), 505/515nm (green), 540/560nm (orange) and 633/660nm (deep red)
  • Includes mounting protocol for user's convenience and sufficient mounting medium to prepare about 100 slides of each

Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)

Order Info

Shipping Condition: Room temperature

Specifications

Type Microscope Point Source Kit
Color Orange, Deep Red, Blue, Green
Format Suspension Beads
Calibration Type Optic Calibration, Fluorescence Microscope Calibration
Content And Storage Store in refrigerator (2–8°C) and protect from light.
For Use With (Application) Internal or External Fluorescence Microscopy Standards
Shipping Condition Room Temperature
Product Line PS-Speck
Quantity 1 kit
What is the refractive index of the mounting medium in PS-Speck Microscope Point Source Kit (Cat. No. P7220)?

The mountant (Component E) has a refractive index of ~1.47.

For Research Use Only. Not for use in diagnostic procedures.