Learn More
Invitrogen™ Multiplexed Proteomics™ Phosphoprotein Gel Staining Kit (Includes MPP33301, 40 μl Standard and S12001)
For distinguishing a low amount of a highly phosphorlyated protein from a higher amount of a less phosphorylated protein
Supplier: Invitrogen™ MPM33306
Description
The Multiplexed Proteomics™ Phosphoprotein Gel Stain Kit contains the Pro-Q™ Diamond phosphoprotein gel stain and the SYPRO™ Ruby protein gel stain. Determining the ratio of the Pro-Q Diamond dye to the SYPRO Ruby dye signal intensities for each band or spot provides a measure of the phosphorylation level normalized to the total amount of protein. Using both stains in combination makes it possible to distinguish a low amount of a highly phosphorlyated protein from a higher amount of a less phosphorylated protein.
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Spécifications
| In-Gel Detection | |
| SYPRO Ruby, Pro-Q Diamond | |
| MPP33301, 40uL Standard and S12001 | |
| 1 kit |
| Fluorescence | |
| Phosphoprotein Gel Staining Kit | |
| PRO-Q | |
| Proteins (Phosphoproteins) |
For Research Use Only. Not for use in diagnostic procedures.