missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pro-Q™ Emerald 488 Glycoprotein Gel and Blot Stain Kit
Description
Pro-Q Emerald 300 and Pro-Q Emerald 488 Glycoprotein Stain Kits provide the most advanced technology available for detection of glycoproteins in gels and on blots. Gel staining is rapid and very sensitive. In less than three hours, it is possible to detect as little as 300 pg of glycoprotein per band, depending on the degree of glycosylation.
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Detection Location | In-Blot Detection, In-Gel Detection |
| Detection Method | Fluorescence |
| Label or Dye | Pro-Q Emerald 488 |
| Quantity | 1 kit |
| Shipping Condition | Room Temperature |
| Target Molecule | Proteins (Glycoproteins) |
| Product Line | PRO-Q |
| Product Type | Glycoprotein Staining Kit |
For Research Use Only. Not for use in diagnostic procedures.