missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pro-Q™ Emerald 300 Glycoprotein Gel and Blot Stain Kit

Catalog No. p21857
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21857 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P21857 Supplier Invitrogen™ Supplier No. P21857
Only null left

50-fold more sensitive than standard fuchsin staining

Pro-Q Emerald 300 and Pro-Q Emerald 488 Glycoprotein Stain Kits provide the most advanced technology available for detection of glycoproteins in gels and on blots. Gel staining is rapid and very sensitive. In less than three hours, it is possible to detect as little as 300 pg of glycoprotein per band, depending on the degree of glycosylation.

Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Specifications

Detection Location In-Blot Detection, In-Gel Detection
Detection Method Fluorescence
Label or Dye Pro-Q Emerald 300
Quantity 1 kit
Shipping Condition Room Temperature
Target Molecule Proteins (Glycoproteins)
Product Line PRO-Q
Product Type Glycoprotein Staining Kit

For Research Use Only. Not for use in diagnostic procedures.