missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pro-Q™ Diamond Phosphoprotein Blot Stain Kit

Catalog No. P33356
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P33356 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P33356 Supplier Invitrogen™ Supplier No. P33356
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Can be used to analyze the native phosphorylation state of proteins obtained from tissue specimens or from body fluids

The Pro-Q Diamond Phosphoprotein Blot Stain Kit provides a rapid and simple method for directly detecting phosphoproteins on polyvinylidene difluoride (PVDF) or nitrocellulose membranes. The fluorescent stain binds selectively to the phosphate moiety and binding is independent of the amino acid sequence context.

Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Specifications

Detection Location In-Blot Detection
Detection Method Fluorescence
Label or Dye Pro-Q Diamond
Quantity 1 kit
Target Molecule Proteins (Phosphoproteins)
Product Line PRO-Q
Product Type Phosphoprotein Blot Stain Kit

For Research Use Only. Not for use in diagnostic procedures.