missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Press-to-Seal™ Silicone Isolator with Adhesive, one well, 20 mm diameter, 0.5 mm deep

Catalog No. P24740
Encompass
Change view
Click to view available options
Quantity:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24740 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P24740 Supplier Invitrogen™ Supplier No. P24740

Removable hydrophobic barriers that offer flexibility for isolating specimens

  • Used to isolate cells grown in culture dishes or to separate specimens on microscopy slides during staining procedures
  • Silicone material is autoclavable and adheres to smooth surface

Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunohistochemistry (IHC)

Order Info

Shipping Condition: Room temperature

Specifications

Type Silicone Isolator
Wells 1
Color Red
Surface Treatment Cell Culture
Material Silicone
Thickness (Metric) 0.5 mm
Content And Storage Store at room temperature.
Depth (Metric) 0.5 mm
Diameter (Metric) 20 mm
Shipping Condition Room Temperature
No. per Pack 1 set
Product Line Press-to-Seal
Quantity 1 set
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.