missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PED6 (N-((6-(2,4-Dinitrophenyl)amino)hexanoyl)-2-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoyl)-1-Hexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt)

Catalog No. D23739
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23739 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D23739 Supplier Invitrogen™ Supplier No. D23739

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PED6-labeled phospholipid

Molecular Probes phospholipase substrates are dye-labeled phospholipids used to monitor phospholipase A (PLA) activity in purified enzyme preparations, cell lysates, and living cells.

  • Label (Ex/Em): PED6
  • Specificity: PLA2 (this substrate is cleaved by PLA1, but this reaction does not separate the fluorophore from the quencher)
  • Cleavage product: BODIPY™ FL C5
  • Lyophilized product can be dissolved in DMSO for use
  • Fluorescence is measured either in microplate assays or using flow cytometry
Note: Two structural variations of these substrates are available, which confer specificity for PLA1 and PLA2. Fluorescently labeled versions of the cleavage product generated from these substrates are also available for use as standards for evaluating percentage substrate conversion in enzymatic reactions.

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Room temperature

Specifications

Quantity 1 mg
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Conjugate Other Labels or Dyes
Product Type Phospholipids

For Research Use Only. Not for use in diagnostic procedures.