Learn More
Invitrogen™ Oregon Green™ 488 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine (Oregon Green™ 488 DHPE) DFS Item
With excitation/emission maxima ∽501/526 nm
Supplier:  Invitrogen™ O12650
Catalog No. O12650
Only null left
Explore available promotions
Description
Description
The phospholipid, Oregon Green 488 DHPE is labeled on the head group with the green-fluorescent Oregon Green fluorophore. This fluorinated analog of fluorescein overcomes some of the key limitations of fluorescein, including greater photostability and a lower pKa (pKa ∽4.7 versus 6.4 for fluorescein), making its fluorescence essentially pH insensitive in the physiological pH range.
Cell Analysis, Cell Viability, Proliferation & Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Ionic Homeostasis & Signaling, pH Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
Phospholipids
1 mg
Solid
Oregon Green
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.