missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NPE-caged ATP (Adenosine 5'-Triphosphate, P3-(1-(2-Nitrophenyl)Ethyl) Ester, Disodium Salt)

Catalog No. A1048
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1048 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1048 Supplier Invitrogen™ Supplier No. A1048
  • ONLINE EXCLUSIVE PRICE
    $489.00
    $400.98 / Each

    Save $88.02

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Flash UV photolysis of caging group creates release of free nucleotides

NPE-caged ATP is a nucleotide analog in which the terminal phosphate is esterified with a blocking group, rendering the molecule biologically inactive. Flash photolysis of the caging group with UV illumination at 360nm results in a rapid and highly localized release of the free nucleotide at the site of illumination.

Enzyme and Protein Activity Assays, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet ice

Specifications

Product Type NPE-Caged ATP
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Shipping Condition Wet Ice
Label or Dye NPE (P3-(1-(2-nitrophenyl)ethyl) ester)
Quantity 5 mg

For Research Use Only. Not for use in diagnostic procedures.