missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NeuroTrace™ DiI Tissue-Labeling Paste
Consists of Dil mixed into inert, water-resistant gel
Supplier: Invitrogen™ N22880
Description
- Ready to use as supplied and can be applied directly to live or fixed tissue specimens using the tip of needle
Cell Analysis, Cell Tracing and Tracking, Neuronal Tracing
Order Info
Shipping Condition: Room temperature
Specifications
| Yellow | |
| Tissue Labeling Paste | |
| Store at room temperature and protect from light. | |
| Fluorescence Microscope | |
| NEUROTRACE |
| Fluorescence | |
| 500 mg | |
| Room Temperature | |
| Cell Membranes & Lipids |
For Research Use Only. Not for use in diagnostic procedures.