missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NeuroTrace™ CM-DiI Tissue-Labeling Paste

Catalog No. N22883
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N22883 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N22883 Supplier Invitrogen™ Supplier No. N22883

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Consists of CM-DiI mixed into an inert, water-resistant gel

  • Ready to use as supplied and can be applied directly to live or fixed tissue specimens using tip of needle
  • Improves penetration of dye into bundled neurons, labeling axons both on and below surface

Cell Analysis, Cell Tracing and Tracking, Neuronal Tracing

Order Info

Shipping Condition: Room temperature

Specifications

Color Yellow
Detection Method Fluorescence
Product Type Tissue Labeling Paste
Quantity 100 mg
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope
Sub Cellular Localization Cell Membranes & Lipids
Product Line NEUROTRACE

For Research Use Only. Not for use in diagnostic procedures.