missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NeuroTrace™ 530/615 Red Fluorescent Nissl Stain - Solution in DMSO

Catalog No. N21482
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N21482 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N21482 Supplier Invitrogen™ Supplier No. N21482
  • ONLINE EXCLUSIVE PRICE
    $382.00
    $313.24 / Each

    Save $68.76

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Selective for Nissl substance characteristic of neurons and provides more sensitivity than traditional histological dyes like toluidine blue or cresyl violet

  • Nissl staining is standard histological method for visualizing neurons in brain and spinal cord
  • Composed of ribosomal RNA associated with rough endoplasmic reticulum in neuronal perikarya and dendrites, Nissl substance redistributes within cell body in injured or regenerating neurons, providing marker for physiological state of neuron

Cell Analysis, Cell Structure, Cell Tracing and Tracking, Neuronal Tracing, Nissl Bodies

Order Info

Shipping Condition: Room temperature

Specifications

Color Orange
Detection Method Fluorescence
Product Type Nissl Stain
Quantity 1 mL
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope
Sub Cellular Localization Nissl Bodies
Excitation/Emission 530/615 nm
Product Line NEUROTRACE
I am labeling brain cryosections with a NeuroTrace Nissl Stain. Is this compatible with antibody labeling?

Yes. We have done this successfully with an anti-GFAP primary and an Alexa Fluor secondary antibody. We would recommend labeling with the primary and secondary antibodies first, then following up with the standard NeuroTrace Nissl Stain protocol.

I can use the NeuroTrace Nissl stains for staining glia or other cell types. What can I do to improve the staining so that it is more selective for neurons?

Our NeuroTrace Nissl stains label the Nissl substance which is composed of ribosomal RNA associated with the rough endoplasmic reticulum and is present in high amounts in neuronal cells. These dyes are not completely specific for neurons, but will selectively stain neurons based on their high level of protein synthesis. In some cases they may show staining of other cell types such as glia, so you may need to decrease the staining concentration to obtain more selective neuronal labeling. We suggest dilutions in the range of 20- to 300-fold for neuronal staining.

Can the NeuroTrace Nissl stains be used on paraffin sections?

We have only tested them on mouse brain cryosections, however, there is at least one citation describing their use on paraffin tissue sections (Michelle L. Schlief, Ann Marie Craig, and Jonathan D. Gitlin. NMDA Receptor Activation Mediates Copper Homeostasis in Hippocampal Neurons. The Journal of Neuroscience, January 5, 2005, 25(1):239 - 246).

What are the NeuroTrace Nissl stains?

The dyes are proprietary, however they are stains that label the Nissl substance, which is composed of ribosomal RNA associated with the rough endoplasmic reticulum and is present in high amounts in neuronal cells.

What products do you have for neuronal tracing?

Please check out this web page (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing.html) for details.

For Research Use Only. Not for use in diagnostic procedures.