missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NBD-PE (N-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)-1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt)

Catalog No. n360
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N360 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N360 Supplier Invitrogen™ Supplier No. N360

The phospholipid, NBD-PE is labeled on the head group with the environment sensitive fluorophore, NBD.

The phospholipid, NBD-PE is labeled on the head group with the environment sensitive fluorophore, NBD (excitation/emission maxima ∼463/536 nm).

Prepare stock solutions of phosphoethanolamines in ethanol to attain a stock concentration of 1 to 2 mg/mL. Sonication may be necessary to aid dispersion.

Order Info

Shipping Condition: Room Temperature

Specifications

Chemical Name or Material Phospholipids
Quantity 10 mg
Molecular Weight (g/mol) 956.25
Physical Form Solid
Product Line Invitrogen
Excitation/Emission 463/536 nm

For Research Use Only. Not for use in diagnostic procedures.