missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NBD Cholesterol (22-(N-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)Amino)-23,24-Bisnor-5-Cholen-3β-Ol)

Catalog No. N1148
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1148 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1148 Supplier Invitrogen™ Supplier No. N1148

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This fluorescent analog, NBD cholesterol, is an environment-sensitive probe that localizes in the membrane's interior and is useful for investigating lipid transport processes as well lipid-protein interactions.

This fluorescent analog, NBD cholesterol, is an environment-sensitive probe that localizes in the membrane's interior and is useful for investigating lipid transport processes as well lipid-protein interactions.

We recommend dissolving in chloroform (CHCl3) or acetonitrile (MeCN) to attain a stock concentration up to 1 mM.

Order Info

Shipping Condition: Room Temperature

Specifications

Chemical Name or Material Cholesterol Analogs
Quantity 10 mg
Physical Form Solid
Product Line Invitrogen
Excitation/Emission 469/537 nm

For Research Use Only. Not for use in diagnostic procedures.