missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NBD C6-Ceramide Complexed to BSA

Catalog No. n22651
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N22651 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N22651 Supplier Invitrogen™ Supplier No. N22651

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Selective stain for Golgi apparatus in live and fixed cells

NBD C6-ceramide can be used to study sphingolipid transport and metabolism mechanisms. Complexing fluorescent lipids with bovine serum albumin (BSA) facilitates cell labeling by eliminating the need for organic solvents to dissolve the lipophilic probe - the BSA-complexed probe can be directly dissolved in water. The environment sensitive NBD dye is weakly fluorescent in water but increases in aprotic solvents and other nonpolar environments with excitation/emission maxima ∽466/536nm.

Cell Analysis, Cell Structure, Golgi

Order Info

Shipping Condition: Room Temperature

Specifications

Chemical Name or Material Sphingolipids
Quantity 5 mg
Physical Form Solid
Product Line Invitrogen

For Research Use Only. Not for use in diagnostic procedures.