Learn More
Invitrogen™ NBD C6-Ceramide (6-((N-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)amino)hexanoyl)Sphingosine)
Selective stain for Golgi apparatus in live and fixed cells
Supplier: Invitrogen™ N1154
Description
NBD C6-ceramide can be used to study sphingolipid transport and metabolism mechanisms. The environment sensitive NBD dye is weakly fluorescent in water but increases in aprotic solvents and other nonpolar environments with excitation/emission maxima ∽466/536nm.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Spécifications
| Sphingolipids | |
| 1 mg | |
| Solid | |
| Invitrogen |
For Research Use Only. Not for use in diagnostic procedures.