missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Octadecyl Rhodamine B Chloride (R18)

Catalog No. o246
Encompass
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
Catalog No. Quantity
O246 10 mg
1 options

Catalog No. O246

Supplier: Invitrogen™ O246

Only null left

Lipophilic octadecyl rhodamine B binds to membranes with fluorophore at aqueous interface and alkyl tail protruding into lipid interior

Cell Analysis, Cell Structure, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Membranes (General) & Lipids, Plasma Membrane

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 10 mg
Shipping Condition Room Temperature
Sub Cellular Localization Cell Membranes & Lipids
Product Line Molecular Probes
Molecular Formula C46H67ClN2O3
Physical Form Solid
Product Type Cell Labeling Reagent

For Research Use Only. Not for use in diagnostic procedures.