missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Measure-IT™ High-Sensitivity Nitrite Assay Kit

Catalog No. M36051
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M36051 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M36051 Supplier Invitrogen™ Supplier No. M36051

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides add-and-read convenience and high-throughput scalability to quantitation of nitrite, thiol, and lead/cadmium

Measure-iT™ assay kits bring add-and-read convenience and high-throughput scalability to the quantitation of nitrite, thiol, and lead/cadmium. The Measure-iT™ High-Sensitivity Nitrite Assay Kit features solid reproducibility over a wide quantitation range, as well as excellent tolerance of common sample contaminants. The assay is linear from 20 to 500 pmoles of nitrite, making it up to 50 times more sensitive than colorimetric assays based on the Griess reagent. The kit includes a nitrite quantitation standard and a protocol that's optimized for microplate-based application.

For Research Use Only. Not for use in diagnostic procedures.

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 1 Kit
For Use With (Application) Nitrite Quantitation
Detection Method Fluorescence
Product Line Measure-IT
Product Type Nitrite Assay Kit
Excitation Wavelength Range 365 nm
Emission Wavelength Range 450 nm

For Research Use Only. Not for use in diagnostic procedures.