missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Marina Blue™ 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine (Marina Blue™ DHPE)

Catalog No. m12652
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12652 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12652 Supplier Invitrogen™ Supplier No. M12652

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A phospholipid

Marina Blue DHPE is labeled on the head group with the blue-fluorescent Marina Blue fluorophore with excitation/emission maxima ∽365/460nm.

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Room Temperature

Specifications

Chemical Name or Material Phospholipids
Quantity 1 mg
Physical Form Solid
Product Line MARINA BLUE

For Research Use Only. Not for use in diagnostic procedures.